missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfinic acid sodium salt, 98%

Product Code. 11480053
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480053 500 g 500g
11460053 25 g 25g
11470053 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480053 Supplier Thermo Scientific Alfa Aesar Supplier No. A12118.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-55-2 | C6H5NaO2S | 164.154 g/mol

It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidants. Store in cool, dry place in a tightly sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-55-2
Molecular Formula C6H5NaO2S
Molecular Weight (g/mol) 164.154
MDL Number MFCD00013135
InChI Key CHLCPTJLUJHDBO-UHFFFAOYSA-M
Synonym sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate
PubChem CID 2723840
IUPAC Name sodium;benzenesulfinate
SMILES C1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 500 g
Beilstein 3598405
Solubility Information Soluble in water.
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material Benzenesulfinic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.