missing translation for 'onlineSavingsMsg'
Learn More

Benzenebutanoic Acid Ethyl Ester, TRC

Product Code. 30340398
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340398 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340398 Supplier Toronto Research Chemicals Supplier No. TRCB1852901G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzenebutanoic Acid Ethyl Ester
CAS 10031-93-3
Molecular Formula C12H16O2
Quantity 1 g
Synonym Ethyl 4-phenylbutanoate, Ethyl 4-phenylbutyrate, NSC 163318
InChI Formula InChI=1S/C12H16O2/c1-2-14-12(13)10-6-9-11-7-4-3-5-8-11/h3-5,7-8H,2,6,9-10H2,1H3
SMILES CCOC(=O)CCCc1ccccc1
IUPAC Name ethyl 4-phenylbutanoate
Molecular Weight (g/mol) 192.25
Formula Weight 192.115
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.