missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzene-1,2,3-triol (Pyrogallol), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Benzene-1,2,3-triol (Pyrogallol)
Specifications
Specifications
| Chemical Name or Material | Pyrogallol |
| CAS | 87-66-1 |
| Analyte or Component Names | Pyrogallol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H6 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1,2,3-Benzenetriol, Pyrogallol (8CI), 1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, 2,6-Dihydroxyphenol, Antioxidant PY, Benzene-1,2,3-triol, C.I. 76515, C.I. Oxidation Base 32, Fouramine Brown AP, Fourrine 85, Fourrine PG, Gallol, NSC 5035, Pyrogallic acid, Pyrogallol acid, Phloroglucinol Anhydrous Imp. A (EP), Phloroglucinol Impurity A |
| InChI Formula | InChI=1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H |
| SMILES | Oc1cccc(O)c1O |
| IUPAC Name | benzene-1,2,3-triol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction