missing translation for 'onlineSavingsMsg'
Learn More

Benzbromarone, TRC

Product Code. 30133455
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133455 1 g 1g
30132400 5 g 5g
18033930 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133455 Supplier Toronto Research Chemicals Supplier No. TRCB1852001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3562-84-3
Molecular Formula C17 H12 Br2 O3
Molecular Weight (g/mol) 424.08
Synonym Benzbromarone, (3,5-Dibromo-4-hydroxyphenyl)(2-ethyl-3-benzofuranyl)methanone;
IUPAC Name (3,5-dibromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone
SMILES CCc1oc2ccccc2c1C(=O)c3cc(Br)c(O)c(Br)c3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H12Br2O3/c1-2-13-15(10-5-3-4-6-14(10)22-13)16(20)9-7-11(18)17(21)12(19)8-9/h3-8,21H,2H2,1H3
Formula Weight 421.9153
Percent Purity >95%
Chemical Name or Material Benzbromarone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.