missing translation for 'onlineSavingsMsg'
Learn More

Benzbromarone, MedChemExpress

Product Code. 16438636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438636 Supplier Medchem Express Supplier No. HYB1135
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benzbromarone is a highly effective and well tolerated non-competitive inhibitor of xanthine oxidase, used as an uricosuric agent, used in the treatment of gout.

Benzbromarone is a highly effective and well tolerated non-competitive inhibitor of xanthine oxidase, used as an uricosuric agent, used in the treatment of gout.

Specifications

Chemical Name or Material Benzbromarone
CAS 3562-84-3
Color White
Purity Grade Notes Research
Molecular Formula C17H12Br2O3
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (235.80 mM)
SMILES O=C(C1=CC(Br)=C(O)C(Br)=C1)C2=C(CC)OC3=CC=CC=C23
Molecular Weight (g/mol) 424.08
Formula Weight 424.08
Percent Purity 99.38%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.