missing translation for 'onlineSavingsMsg'
Learn More

Benzanilide, 98%

Product Code. 10330454 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10330454 50 g Plastic bottle 50g
10396450 5 g Glass bottle 5g
10356500 250 g Plastic bottle 250g
10510974 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10330454 Supplier Thermo Scientific Acros Supplier No. 148550500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-98-1 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-98-1
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00003069
InChI Key ZVSKZLHKADLHSD-UHFFFAOYSA-N
Synonym benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline
PubChem CID 7168
IUPAC Name N-phenylbenzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 161°C to 165°C
Color Gray to Green-Yellow
Boiling Point 117°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CONHC6H5
Quantity 50 g
Beilstein 12, 262
Merck Index 15, 1063
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/60 mL alcohol,1g/7 mL boiling alcohol,slightly soluble in ether,acetic acid
Formula Weight 197.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzanilide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.