missing translation for 'onlineSavingsMsg'
Learn More

Benzamil hydrochloride, MedChemExpress

Product Code. 16494607
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494607 50 mg 50mg
16478708 10 mg 10mg
16458786 100 mg 100mg
16401128 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494607 Supplier Medchem Express Supplier No. HYB1546A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benzamil hydrochloride (Benzylamiloride hydrochloride), an Amiloride analogue, is a Na+/Ca2+ exchanger (NCX) inhibitor (IC50∼100 nM). Benzamil hydrochloride also is a non-selective Deg/epithelial sodium channels (ENaC) blocker, and can potentiate myogenic vasoconstriction. Benzamil hydrochloride inhibits TRPP3-mediated Ca2+-activated currents, with an IC50 of 1.1 μM.

Benzamil hydrochloride (Benzylamiloride hydrochloride), an Amiloride analogue, is a Na+/Ca2+ exchanger (NCX) inhibitor (IC50∼100 nM). Benzamil hydrochloride also is a non-selective Deg/epithelial sodium channels (ENaC) blocker, and can potentiate myogenic vasoconstriction. Benzamil hydrochloride inhibits TRPP3-mediated Ca2+-activated currents, with an IC50 of 1.1 μM.

Specifications

Chemical Name or Material Benzamil hydrochloride
CAS 161804-20-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H15Cl2N7O
Quantity 50 mg
Synonym Benzylamiloride hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (280.73 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=NC(Cl)=C(N)N=C1N)NC(NCC2=CC=CC=C2)=N.[H]Cl
Molecular Weight (g/mol) 356.21
Formula Weight 356.21
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.