missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine hydrochloride hydrate, 99%, water ca 10-14%

Product Code. p-7042524
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 15460267

Brand: Thermo Scientific Alfa Aesar J62823.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases | CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol

Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206752-36-5
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
SMILES Cl.NC(=N)C1=CC=CC=C1

Specifications

Melting Point 80°C to 85°C
Color White
Quantity 5 g
Beilstein 3594959
Formula Weight 156.62
Percent Purity 99%
Physical Form Beads or Powder
Assay water ≈10 to 14%
Chemical Name or Material Benzamidine hydrochloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.