missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde, TRC

Product Code. 30338131
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338131 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338131 Supplier Toronto Research Chemicals Supplier No. TRCB119740100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzaldehyde
CAS 100-52-7
Purity Grade Notes GC
Molecular Formula C7 H6 O
Quantity 100 g
Synonym Artificial Almond Oil, Benzaldehyde FFC, Benzenecarbonal, Benzenecarboxaldehyde, Benzoic acid aldehyde, Benzoic aldehyde, NSC 7917, Phenylformaldehyde, Phenylmethanal, Amfetamine Sulfate Impurity D, Amfetamine Impurity D, Benzalkonium Chloride Impurity B, Benzalkonium Impurity B, Benzoyl Peroxide Impurity A, Benzyl Alcohol Impurity A, Dexamfetamine Sulfate Impurity D, Dexamfetamine Impurity D, Fentanyl Citrate Impurity E, Fentanyl Impurity E, Glycopyrronium Bromide Impurity F, Glycopyrronium Impurity F
InChI Formula InChI=1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H
SMILES O=Cc1ccccc1
IUPAC Name benzaldehyde
Molecular Weight (g/mol) 106.12
Formula Weight 106.0419
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.