missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde dimethyl acetal, 99%

Product Code. 11484473
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484473 2500 g 2500g
11464473 100 g 100g
11474473 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484473 Supplier Thermo Scientific Alfa Aesar Supplier No. A11009.0E
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1125-88-8 | C9H12O2 | 152.193 g/mol

Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-88-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00008491
InChI Key HEVMDQBCAHEHDY-UHFFFAOYSA-N
Synonym benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
PubChem CID 62375
IUPAC Name dimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)OC

Specifications

Density 1.014
Boiling Point 196°C
Flash Point 69°C (156°F)
Odor Characteristic
Refractive Index 1.4935
Quantity 2500 g
Beilstein 2044501
Solubility Information Insoluble in water. Soluble in alcohol.
Formula Weight 152.2
Percent Purity 99%
Chemical Name or Material Benzaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.