missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | Colorless |
Density | 1.0100g/mL |
Boiling Point | 87.0°C to 89.0°C (18.0 mmHg) |
Flash Point | 69°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C9H12O2 |
Show More |
Safety and Handling
Product Identifier
- Benzaldehyde dimethyl acetal
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'