missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, MP Biomedicals™

Product Code. 11374099 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374099 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11374099 Supplier MP Biomedicals Supplier No. 0210089410
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Emulsifier, Adsorbent
TRUSTED_SUSTAINABILITY

Specifications

CAS 1302-78-9
Color Gray
Density 2 to 3g/cm3
Molecular Formula Al2O3.4SiO2.2H2O
Quantity 10 kg
Synonym Montmorillonite
Physical Form Solid
Chemical Name or Material Bentonite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.