Learn More
Lenz Laborglasinstrumente™ Bent Splash Heads
Click to view available options
Standard Taper Joint:
NS 29/32 Vertical, NS 29/32 Bent Cone
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | For separation of drops (Direct placement onto distillation flasks) |
| Type | Bent Splash Heads |
| Material | Duran Glass |
| Standard Taper Joint | NS 29/32 Vertical, NS 29/32 Bent Cone |