Learn More
Lenz Laborglasinstrumente™ Bent Receiver Adapter with Glass Olive
Allows for the connection of a condenser to a receiving flask. Lenz Laborglasinstrumente™ Bent Receiver Adapters with Glass Olive can be directly heated and withstand typical laboratory thermal variations.
£30.53 - £36.68
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
11886154
|
Lenz Laborglasinstrumente™
05275002 |
NS 19/26 |
£31.22
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11806134
|
Lenz Laborglasinstrumente™
05275001 |
NS 14/23 |
£30.53
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11866204
|
Lenz Laborglasinstrumente™
05275003 |
NS 24/29 |
£36.36
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11876194
|
Lenz Laborglasinstrumente™
05275004 |
NS 29/32 |
£36.68
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- NS socket and NS cone
- 105° angle
- Vacuum connection
- Glass olive with drain tip
- Meets DIN 12594 standards
Specifications
Bent Receiver Adapter with Glass Olive | |
Duran Glass |