missing translation for 'onlineSavingsMsg'
Learn More

Benserazide Hydrochloride, TRC

Product Code. 30135540
Change view
Click to view available options
Quantity:
10 mg
100 mg
5 g
Unit Size:
100mg
10mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135540 100 mg 100mg
30135898 5 g 5g
17944945 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135540 Supplier Toronto Research Chemicals Supplier No. TRCB120500100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14919-77-8
Molecular Formula C10 H15 N3 O5 . Cl H
Molecular Weight (g/mol) 293.7
Synonym Benserazide hydrochloride, (2RS)-2-Amino-3-hydroxy-2'-(2,3,4-trihydroxybenzyl)propanohydrazide hydrochloride
IUPAC Name 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride
SMILES Cl.NC(CO)C(=O)NNCc1ccc(O)c(O)c1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H15N3O5.ClH/c11-6(4-14)10(18)13-12-3-5-1-2-7(15)9(17)8(5)16;/h1-2,6,12,14-17H,3-4,11H2,(H,13,18);1H
Formula Weight 293.0778
Chemical Name or Material Benserazide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.