missing translation for 'onlineSavingsMsg'
Learn More

Benserazide hydrochloride, Thermo Scientific™

Inhibitor of L-aromatic amino acid decarboxylase

Brand:  Thermo Scientific Alfa Aesar J64581.03

Quantity 1g

 View more versions of this product

Product Code. 15486469

  • £57.60 / 1g
 Restricted Item
Estimated Shipment: 29-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Inhibitor of L-aromatic amino acid decarboxylaseBenserazide hydrochloride is used as an antiparkinsonian. It is a peripheral decarboxylase inhibitor. It is used as an inhibitor for aromatic L-aminoacid decarboxylase. Further, it serves as an inhibitor of tyrosine hydroxylase (TH) and Tryptophan hydroxylase (TPH) inhibitor.

Solubility
Soluble in water, dimethyl sulfoxide and methanol. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

14919-77-8
293.704
ULFCBIUXQQYDEI-UHFFFAOYSA-N
26964
2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride
C10H16ClN3O5
MFCD00078571
benserazide hydrochloride, benserazide hcl, 2-amino-3-hydroxy-n'-2,3,4-trihydroxybenzyl propanehydrazide hydrochloride, ccris 5092, 2'-2,3,4-trihydroxybenzyl-dl-serinohydrazide monohydrochloride, benzerazide hydrochloride, dl-serine 2, dl-serine 2-2,3,4-trihydroxybenzyl hydrazine hydrochloride, 2-amino-3-hydroxy-n'-2,3,4-trihydroxyphenyl methyl propanehydrazide hydrochloride
CHEBI:31262
C1=CC(=C(C(=C1CNNC(=O)C(CO)N)O)O)O.Cl

Specifications

14919-77-8
MFCD00078571
Light sensitive
benserazide hydrochloride, benserazide hcl, 2-amino-3-hydroxy-n'-2,3,4-trihydroxybenzyl propanehydrazide hydrochloride, ccris 5092, 2'-2,3,4-trihydroxybenzyl-dl-serinohydrazide monohydrochloride, benzerazide hydrochloride, dl-serine 2, dl-serine 2-2,3,4-trihydroxybenzyl hydrazine hydrochloride, 2-amino-3-hydroxy-n'-2,3,4-trihydroxyphenyl methyl propanehydrazide hydrochloride
ULFCBIUXQQYDEI-UHFFFAOYSA-N
2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride
26964
293.7
C10H16ClN3O5
1g
14,1050
Soluble in water,dimethyl sulfoxide and methanol. Insoluble in ethanol and acetone.
C1=CC(=C(C(=C1CNNC(=O)C(CO)N)O)O)O.Cl
293.704
CHEBI:31262
Benserazide hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.