missing translation for 'onlineSavingsMsg'
Learn More

Benserazide hydrochloride, MedChemExpress

Product Code. 16442159
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442159 1 g 1g
16410918 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442159 Supplier Medchem Express Supplier No. HYB0404A
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benserazide hydrochloride (Serazide) is commonly used in Parkinson's disease and is an inhibitor of peripheral aromatic L-amino acid decarboxylase (AADC).

Benserazide hydrochloride (Serazide) is commonly used in Parkinson's disease and is an inhibitor of peripheral aromatic L-amino acid decarboxylase (AADC).

Specifications

Chemical Name or Material Benserazide hydrochloride
CAS 14919-77-8
Color White
Purity Grade Notes Research
Molecular Formula C10H16ClN3O5
Quantity 1 g
Synonym Serazide Ro 4-4602
Solubility Information DMSO : 100 mg/mL (340.48 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (170.24 mM)
SMILES NC(CO)C(NNCC1=CC=C(O)C(O)=C1O)=O.Cl
Molecular Weight (g/mol) 293.7
Formula Weight 293.7
Percent Purity 97.4%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.