missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benorilate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Benorilate |
| CAS | 5003-48-5 |
| Molecular Formula | C17 H15 N O5 |
| Quantity | 1 g |
| Synonym | Salicylic acid acetate, ester with 4'-hydroxyacetanilide (7CI,8CI), Acetanilide, 4'-hydroxy-, salicylate (ester) acetate (ester) (8CI), 4-Acetamidophenyl 2-acetoxybenzoate, 4-Acetaminophenyl 2-acetoxybenzoate, Acetylsalicylic acid paracetamol ester, Aspirin acetaminophen ester, Benoral, Benorilate, Benortan, Benorylate, Fenasprate, Quinexin, Salipran, TO 125, Win 11450, p-Acetamidophenyl acetylsalicylate, p-N-Acetylaminophenyl acetylsalicylate, Benzoic acid, 2-(acetyloxy)-, 4-(acetylamino)phenyl ester |
| InChI Formula | InChI=1S/C17H15NO5/c1-11(19)18-13-7-9-14(10-8-13)23-17(21)15-5-3-4-6-16(15)22-12(2)20/h3-10H,1-2H3,(H,18,19) |
| SMILES | CC(=O)Nc1ccc(OC(=O)c2ccccc2OC(=O)C)cc1 |
| IUPAC Name | (4-acetamidophenyl) 2-acetyloxybenzoate |
| Molecular Weight (g/mol) | 313.3 |
| Formula Weight | 313.095 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction