missing translation for 'onlineSavingsMsg'
Learn More

Bendazac, TRC

Product Code. 30329062
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329062 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329062 Supplier Toronto Research Chemicals Supplier No. TRCB1326002G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bendazac
CAS 20187-55-7
Molecular Formula C16H14N2O3
Quantity 2 g
Synonym [[1-(Phenylmethyl)-1H-indazol-3-yl]oxy]acetic Acid, 1-Benzylindazole-3-oxyacetic Acid, [(1-Benzyl-1H-indazol-3-yl)oxy]acetic Acid, AF 983, Bendazolic Acid, Bindazac, Dogalina, Zildasac, Zildazac
InChI Formula InChI=1S/C16H14N2O3/c19-15(20)11-21-16-13-8-4-5-9-14(13)18(17-16)10-12-6-2-1-3-7-12/h1-9H,10-11H2,(H,19,20)
SMILES OC(=O)COc1nn(Cc2ccccc2)c3ccccc13
IUPAC Name 2-(1-benzylindazol-3-yl)oxyacetic acid
Molecular Weight (g/mol) 282.29
Formula Weight 282.1004
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.