missing translation for 'onlineSavingsMsg'
Learn More

Bendazac, Thermo Scientific Chemicals

Product Code. 16594444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16594444 5 g 5g
16584444 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594444 Supplier Thermo Scientific Acros Supplier No. 464920050
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20187-55-7
Molecular Formula C16H13N2NaO3
Molecular Weight (g/mol) 304.28
InChI Key GHFGHIKJGVMEFT-UHFFFAOYSA-M
IUPAC Name sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate
SMILES [Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 160.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform, ethanol, acetone
Formula Weight 282.3
Chemical Name or Material Bendazac

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.