missing translation for 'onlineSavingsMsg'
Learn More

Bendamustine Hydrochloride, TRC

Product Code. 30341099
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341099 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341099 Supplier Toronto Research Chemicals Supplier No. TRCB13250025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bendamustine Hydrochloride
CAS 3543-75-7
Molecular Formula C16H21Cl2N3O2.HCl
Quantity 25 mg
Synonym 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, hydrochloride (1:1), 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, monohydrochloride (9CI), 2-Benzimidazolebutyric acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, monohydrochloride (8CI), Bendamustin hydrochloride, Bendamustine hydrochloride, Bendit, Cytostasan, IMET 3393, Purplz, Ribomustin, SDX 105, Treanda, ZIMET 33/93
InChI Formula InChI=1S/C16H21Cl2N3O2.ClH/c1-20-14-6-5-12(21(9-7-17)10-8-18)11-13(14)19-15(20)3-2-4-16(22)23;/h5-6,11H,2-4,7-10H2,1H3,(H,22,23);1H
SMILES Cl.Cn1c(CCCC(=O)O)nc2cc(ccc12)N(CCCl)CCCl
IUPAC Name 4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid;hydrochloride
Molecular Weight (g/mol) 394.72
Formula Weight 393.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.