missing translation for 'onlineSavingsMsg'
Learn More

Bendamustine hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18752784
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18752784 50 mg 50mg
18702804 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752784 Supplier Tocris Bioscience Supplier No. 3681/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cytostatic agent; exhibits DNA alkylating and purine analog properties

Cytostatic agent that displays activity in non-Hodgkin's lymphomas. Exhibits bifunctionality; combines DNA alkylating properties with those of purine analogs.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3543-75-7
Molecular Formula C16H22Cl3N3O2
Molecular Weight (g/mol) 394.721
InChI Key ZHSKUOZOLHMKEA-UHFFFAOYSA-N
Synonym bendamustine hydrochloride, treanda, bendamustine hcl, ribomustin, cytostasan, bendamustin hydrochloride, treakisym, 4-5-bis 2-chloroethyl amino-1-methyl-1h-benzo d imidazol-2-yl butanoic acid hydrochloride, bendeka
PubChem CID 77082
IUPAC Name 4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid;hydrochloride
SMILES CN1C2=C(C=C(C=C2)N(CCCl)CCCl)N=C1CCCC(=O)O.Cl

Specifications

Quantity 50 mg
Formula Weight 394.72
Percent Purity >98%
Chemical Name or Material Bendamustine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.