Learn More
Gibco™ BenchStable™ DMEM

Description
Gibco DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
BenchStable DMEM has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional DMEM when supplemented with ≥10% fetal bovine serum (FBS).
This DMEM is modified as follows:
• Optimized for room temperature stability
• With High Glucose, GlutaMAX, Phenol Red
• Without Sodium Pyruvate
Using BenchStable DMEM
DMEM is unique from other media as it contains four times the concentration of amino acids and vitamins as the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and so requires a 5–10% CO2 environment to maintain physiological pH. DMEM with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable DMEM contains no proteins, lipids, or growth factors and so requires supplementation with ≥10% fetal bovine serum (FBS).
Specifications
Specifications
| Cell Line | HeLa, 293, Cos-7, and PC-12 |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| With Additives | High Glucose, GlutaMAX, Phenol Red |
| Without Additives | No Sodium Pyruvate, No HEPES |
| Manufacturing Quality | cGMP-compliant under the ISO 9001: 2015 standard |
| Show More |
Frequently Asked Questions (FAQs)
We recommended supplementing BenchStable media with at least 10% FBS for best results. If your culture method calls for greater than 10% FBS supplementation using our other existing media, we recommend that you use the same concentration with BenchStable media.
We recommend supplementing BenchStable media with at least 10% FBS for best results. We have run tests on limited cell types using 5% FBS concentration and have not observed compromised performance. Please contact Tech Support at techsupport@thermofisher.com to ensure that your specific application allows for 5% serum supplementation.
We have tested our BenchStable media for growth and morphology on the following cell lines: A549, CHO-K1, HEK293, HeLa, HL-60, Jurkat, MCF7, MDA-MB-231, Neuro2A, SH-SY5Y, and THP-1.
At this time, we do not offer BenchStable media without phenol red.
The shelf life of BenchStable media is unchanged when stored cold (2-4 degrees C) or ambient (15-30 degrees C).
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.