Learn More
Invitrogen™ BenchPro™ 2100 Waste Tray
Product Code. 11380972
Change view
Click to view available options
Quantity:
2 Packs
missing translation for 'unitSize'
Each
Product Code. Quantity unitSize
11380972 2 Packs Each
1 missing translation for 'options'
Product Code. 11380972 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' MC4001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
BenchPro™ 2100 Waste Tray
The BenchPro™ 2100 Waste Tray is used with the BenchPro™ 2100 Plasmid Processing Station (Cat# MC1001) and supports the disposable Reagent Tray inside the instrument drawer.
TRUSTED_SUSTAINABILITY
Specifications
Product Type BenchPro™ 2100 Waste Tray
Description BenchPro™ 2100 Waste Tray, 1 Pack of 2, Tray Type, Store at Room Temperature, Used With BenchPro™ 2100 Plasmid Purification System, Ideal for Automated Nucleic Acid Purification, DNA Extraction
For Use With (Equipment) BenchPro™ 2100 plasmid purification system
Includes 1 Pack of 2
Quantity 2 Packs