Learn More
Specac™ Benchmark™ Series Laser Alignment Jig Accessory
Product Code. 15250328
Encompass_Preferred
Click to view available options
Type:
Benchmark Series Laser Alignment Jig Accessory
Product Code. 15250328
Brand: Specac™ GS24500
This accessory allows the visual verification of the optical pathlength through the Cyclone series gas cells. This is very useful when different pathlengths are used with variable pathlength cells.
Specifications
Type Benchmark Series Laser Alignment Jig Accessory