missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Bench Top Timer with External Triggering Contacts

Product Code. 17495303
Click to view available options
Time Range:
9 hr. 59 min. 59 sec.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17495303

Brand: Brannan™ 28/410/0

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Brannan bench top timer is easy to use with color coded buttons and a clear LCD display designed to offer accurate measurement over a long period of time.

  • Three 4 mm sockets for external triggering
  • 1 m cable
  • Large LCD and buttons
  • Up to 9 hours, 59 minutes, 59 seconds
  • 1/100 second resolution
  • Includes batteries and operating instructions
  • 0 to 50°C

Specifications

Product Type Benchtop Timer
Time Range 9 hr. 59 min. 59 sec.
Accuracy ± 1 sec.
Resolution 1/100 sec.
Display LCD
Increments Seconds
Battery Type 1 x AA
Color Black
Weight (English) 0.8708 lb.
Weight (Metric) 0.395 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.