missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Bench Top pH Meters
Click to view available options
Unit Size:
Each
Accuracy:
±0.01
Description
A low cost, space efficient bench top pH meter, with a built-in magnetic stirrer. The HI-208 also features automatic pH calibration, automatic temperature compensation and is supplied complete with pH electrode which incorporates the temperature sensor. Powered by mains electricity or the supplied 9V battery (run time up to 500hr).
- Compact bench pH meter
- Built-in magnetic stirrer
- Mains or batteries operated
- Built-in sample beaker and electrode holder
- Dual level LCD display for pH and temperature
- pH electrode with temperature sensor included
- Automatic pH calibration and temperature compensation
Specifications
Specifications
| Description | Bench Top pH Meter |
| Accuracy (Temperature) | ±0.5°C |
| Accuracy (pH) | ±0.02 |
| For Use With (Application) | For Education |
| Calibration | Automatic |
| Range (pH) | -2 to 16 |
| Resolution (pH) | 0.01 |
| Calibration Points | 1 or 2 with 2 sets of memorized buffers |
| Temperature Range (Metric) | -5.0°C to +105.0°C |
| Resolution (Temperature) | 0.1°C |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido