missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ Bench-Top Forced Convection Chamber Furnace

Product Code. p-7236262 Shop All Nabertherm Products
Click to view available options
Capacity (Metric):
15 L
50 L
Controller Type:
B510
C550
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17358069

Brand: Nabertherm™ NAT0181L1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

These chamber furnaces with air circulation are characterized by their extremely high temperature uniformity. Suitable for processes such as annealing, crystalizing, preheating, curing, but also for numerous processes in tool making.

  • Max temp 850°C
  • Horizontal air circulation with optimum distribution through stainless steel baffles
  • Swing door hinged on the right

Specifications

Controller Type B510
Capacity (Metric) 15 L
Temperature (Metric) 850°C
Type Furnace
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.