Learn More
Kern Sohn Bench Scale FKB
High-resolution bench scale with large weighing range and robust stainless steel weighing plate.
£335.35 - £479.06
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
17391633
|
Kern Sohn
FKB 15K0.5 |
15 kg | 0.5 g |
£335.35
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17321643
|
Kern Sohn
FKB 16K0.05 |
16 kg | 0.05 g |
£479.06
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17331643
|
Kern Sohn
FKB 16K0.1 |
16 kg | 0.1 g |
£431.15
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17341643
|
Kern Sohn
FKB 30K1 |
30 kg | 1 g |
£339.70
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17361643
|
Kern Sohn
FKB 36K0.1 |
36 kg | 0.1 g |
£479.06
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17371643
|
Kern Sohn
FKB 36K0.2 |
36 kg | 0.2 g |
£479.06
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17381643
|
Kern Sohn
FKB 65K0.2 |
65 kg | 0.2 g |
£457.29
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17321653
|
Kern Sohn
FKB 6K0.02 |
6 kg | 0.02 g |
£453.42
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17331653
|
Kern Sohn
FKB 8K0.05 |
8 kg | 0.05 g |
£418.09
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17341653
|
Kern Sohn
FKB 8K0.1 |
8 kg | 0.1 g |
£339.70
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Thanks to the high resolution it is ideal for high-precision weighing in the industrial field
- Housing with robust aluminum diecast housing trough, maintains the stability, protects the weighing technology elements and is robust enough to cope with everyday use
- Particularly practical: thanks to the large weighing ranges and compact dimensions, you can accurately weigh heavy loads in the most restricted of spaces. Useful for determining very small weight differences, such as, for example, gas wastage, abrasion of mechanical parts, rock samples, minerals, druses, silver etc.
- PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
- Freely programmable weighing unit, e.g. display direct in special units such as length of wire g/m, surface weight g/m2, or else
- Protective working cover included with delivery
Specifications
Stainless Steel |