missing translation for 'onlineSavingsMsg'
Learn More

Benazeprilat, TRC

Product Code. 30132871
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132871 5 mg 5mg
17944775 10 mg 10mg
17934625 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132871 Supplier Toronto Research Chemicals Supplier No. TRCB1197605MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86541-78-8
Molecular Formula C22 H24 N2 O5
Molecular Weight (g/mol) 396.44
Synonym 1H-1-Benzazepine-1-acetic acid, 3-[[(1S)-1-carboxy-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, (3S)-;1H-1-Benzazepine-1-acetic acid, 3-[(1-carboxy-3-phenylpropyl)amino]-2,3,4,5-tetrahydro-2-oxo-, [S-(R*,R*)]-, Benazeprilat, Benazeprilate, CGS 14831, Benazepril Related Compound C (USP), Benazepril Hydrochloride Imp. C (EP)
IUPAC Name (2S)-2-[[(3S)-1-(carboxymethyl)-2-oxo-4,5-dihydro-3H-1-benzazepin-3-yl]amino]-4-phenylbutanoic acid
SMILES OC(=O)CN1C(=O)[C@H](CCc2ccccc12)N[C@@H](CCc3ccccc3)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H24N2O5/c25-20(26)14-24-19-9-5-4-8-16(19)11-13-17(21(24)27)23-18(22(28)29)12-10-15-6-2-1-3-7-15/h1-9,17-18,23H,10-14H2,(H,25,26)(H,28,29)/t17-,18-/m0/s1
Formula Weight 396.1685
Percent Purity >95%
Chemical Name or Material Benazeprilat
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.