missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benazeprilat, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Benazeprilat |
| CAS | 86541-78-8 |
| Analyte or Component Names | Benazeprilat |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H24 N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benazepril Hydrochloride Imp. C (EP), Benazepril Imp. C (EP), Benazepril USP Related Compound C, Benazepril USP RC C, (2S)-2-[[(3S)-1-(Carboxymethyl)-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl]amino]-4-phenylbutanoic Acid (Benazeprilat), 1H-1-Benzazepine-1-acetic acid, 3-[[(1S)-1-carboxy-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, (3S)-, 1H-1-Benzazepine-1-acetic acid, 3-[(1-carboxy-3-phenylpropyl)amino]-2,3,4,5-tetrahydro-2-oxo-, [S-(R*,R*)]-, Benazeprilat, Benazeprilate, CGS 14831, Benazepril Related Compound C (USP), Benazepril Related Compound C, Benazepril Hydrochloride Impurity C, Benazepril Impurity C |
| InChI Formula | InChI=1S/C22H24N2O5/c25-20(26)14-24-19-9-5-4-8-16(19)11-13-17(21(24)27)23-18(22(28)29)12-10-15-6-2-1-3-7-15/h1-9,17-18,23H,10-14H2,(H,25,26)(H,28,29)/t17-,18-/m0/s1 |
| Solution Type | Neat |
| SMILES | OC(=O)CN1C(=O)[C@H](CCc2ccccc12)N[C@@H](CCc3ccccc3)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?