missing translation for 'onlineSavingsMsg'
Learn More

Benazepril hydrochloride, MedChemExpress

Product Code. 16400778
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16400778

Brand: Medchem Express HYB0093A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Benazepril hydrochloride, an angiotensin converting enzyme inhibitor, which is a medication used to treat high blood pressure.

Benazepril hydrochloride, an angiotensin converting enzyme inhibitor, which is a medication used to treat high blood pressure.

Specifications

Chemical Name or Material Benazepril hydrochloride
Physical Form Solid
CAS 86541-74-4
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C24H29ClN2O5
For Use With (Application) COVID-19-anti-virus
Synonym CGS14824A
Solubility Information DMSO : 100 mg/mL (216.94 mM; Need ultrasonic) ∣H2O : 10 mg/mL (21.69 mM; Need ultrasonic)
SMILES O=C1N(CC(O)=O)C2=CC=CC=C2CC[C@@H]1N[C@@H](CCC3=CC=CC=C3)C(OCC)=O.Cl
Molecular Weight (g/mol) 460.95
Formula Weight 460.95
Percent Purity 99.92%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.