missing translation for 'onlineSavingsMsg'
Learn More

Benazepril Ethyl Ester, TRC

Product Code. 30333451
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333451 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333451 Supplier Toronto Research Chemicals Supplier No. TRCB11981525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benazepril Ethyl Ester
CAS 103129-58-4
Molecular Formula C26 H32 N2 O5
Quantity 25 mg
Synonym 1H-1-Benzazepine-1-acetic acid, 3-[[1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, ethyl ester, [S-(R*,R*)]- (9CI), Benazepril Ethyl Ester, Benazepril Related Compound G (USP), Benazepril Hydrochloride Imp. G (EP)
InChI Formula InChI=1S/C26H32N2O5/c1-3-32-24(29)18-28-23-13-9-8-12-20(23)15-17-21(25(28)30)27-22(26(31)33-4-2)16-14-19-10-6-5-7-11-19/h5-13,21-22,27H,3-4,14-18H2,1-2H3/t21-,22-/m0/s1
SMILES CCOC(=O)CN1C(=O)[C@H](CCc2ccccc12)N[C@@H](CCc3ccccc3)C(=O)OCC
IUPAC Name ethyl (2S)-2-[[(3S)-1-(2-ethoxy-2-oxoethyl)-2-oxo-4,5-dihydro-3H-1-benzazepin-3-yl]amino]-4-phenylbutanoate
Molecular Weight (g/mol) 452.54
Formula Weight 452.2311
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.