missing translation for 'onlineSavingsMsg'
Learn More

Benactyzine Hydrochloride, Mikromol™

Product Code. 18000710
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18000710 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18000710 Supplier Mikromol Supplier No. MM1826.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benactyzine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benactyzine Hydrochloride
CAS 57-37-4
Analyte or Component Names Benactyzine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H25 N O3 . Cl H
Certifications/Compliance ISO 17025
Synonym Benzilic acid, 2-(diethylamino)ethyl ester hydrochloride (6CI,8CI), Ethanol, 2-diethylamino-, benzilate-HCl (4CI), 2-Diethylaminoethyl benzilate hydrochloride, 2-Diethylaminoethyl diphenylglycolate hydrochloride, AY 5406, AY 5406-1, Actozine, Aktozin, Amisyl, Amitakon, Amizil, Amizyl, Amyzyl, Arcadine, Arkadin, Benactyzine chloride, Benactyzine hydrochloride, Benaktin, Cafron, Cedad, Cevanol, Destendo, Fobex, Ibiotyzil, Katron, Leucidil, Lucidil, Nervacton, Nervatil, Neuraktil, Neurobenzil, Neuroleptone, Nutinal, Parasan, Parazan, Parpon, Phobex, Procalm, Prokalm, Stoikon, Suavitil, Tranquillin, Valladan, Valladon, Win 5606, Benzeneacetic acid, alpha-hydroxy-alpha-phenyl-, 2-(diethylamino)ethyl ester, hydrochloride (9CI), beta-Diethylaminoethyl benzilate hydrochloride, Benzeneacetic acid, alpha-hydroxy-alpha-phenyl-, 2-(diethylamino)ethyl ester, hydrochloride (1:1)
InChI Formula InChI=1S/C20H25NO3.ClH/c1-3-21(4-2)15-16-24-19(22)20(23,17-11-7-5-8-12-17)18-13-9-6-10-14-18;/h5-14,23H,3-4,15-16H2,1-2H3;1H
SMILES Cl.CCN(CC)CCOC(=O)C(O)(c1ccccc1)c2ccccc2
IUPAC Name 2-(diethylamino)ethyl 2-hydroxy-2,2-diphenylacetate;hydrochloride
Molecular Weight (g/mol) 363.88
Quantity 100 mg
Formula Weight 363.1601 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.