missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benactyzine hydrochloride, MedChemExpress
Description
Benactyzine hydrochloride is a butyrylcholinesterase (BChE) inhibitor with a Ki of 0.010 mM.
Specifications
Specifications
| Chemical Name or Material | Benactyzine hydrochloride |
| CAS | 57-37-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H26ClNO3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 150 mg/mL (412.22 mM; Need ultrasonic and warming) ∣H2O : 50 mg/mL (137.41 mM; Need ultrasonic) |
| SMILES | O=C(OCCN(CC)CC)C(C1=CC=CC=C1)(O)C2=CC=CC=C2.[H]Cl |
| Molecular Weight (g/mol) | 363.88 |
| Formula Weight | 363.88 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?