Learn More
Emma Safety Footwear Bellagio Safety Shoes
Product Code. 15988231
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15988231
Brand: Emma Safety Footwear MM13709039
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A classically styled Brogue style Business shoe that always sets the right tone. Polished black full grain leather uppers and a virtually impenetrable nitrile rubber sole allows the wearer to stroll from the office to the factory and back again, with no fuss or shoe changing.
  • S3-Safety shoe with sole plate for wet work
  • Nitrile rubber heat resistant outsole
  • Full grain leather construction
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Bellagio
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 39
Toe Material Steel