Learn More
Invitrogen™ Belimumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558447
 View more versions of this product
Product Code. 30282839
  • £397.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Belimumab is an intravenous immunosuppressant for the adjunctive treatment of systemic lupus erythematosus (SLE). More specifically, it is a fully human recombinant IgG1lambda monoclonal antibody produced from a recombinant NS0 cell line stably transfected with the belimumab heavy chain and light chain genes. It is the first biological treatment approved for the indication of SLE. Concomitant use with live or inactivated vaccines must be avoided. Belimumab was FDA approved on March 9, 2011. Belimumab consists of 2 heavy chains, and 2 light chains of the lambda subclass. Each heavy chain contains 452 amino acid residues and each light chain contains 214 amino acid residues. There are 3 post-translational modifications: a conserved N-linked glycosylation on the CH2 domain at Asn 303 of the heavy chain, the conversion of the N-terminal glutamine residue of the heavy chain into pyroglutamate, and loss of C-terminal lysine residue of the heavy chain.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Belimumab
Recombinant Monoclonal
Unconjugated
hBlySmAb-1.1; LymphoStat-B
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized