missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Switch-Grid™ Test Tube Rack Grids

Replacement grids

Brand:  Bel-Art™ F18745-2000

Code : 04

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13442469

  • £5.60 / Pack of 4
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

Bel-Art™ Scienceware™ Switch-Grid™ Test Tube Rack Grids

  • Chemical resistant polyoxymethylene plastic
  • Can be cleaned with mild detergent, ethanol or isopropyl alcohol
  • Autoclavable at121°C (250°F)
Specifications

Specifications

Grid Set
Green
Plastic
5 x 4
Tubes (16 to 20 mm)
40
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers