missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wire Lead Flask with Vikem™ Coating

“Wire” lead weights coil into a helix, spiral, cone or any irregular shape to stabilize vessels in a water bath

Brand:  Bel-Art™ F18324-0001

Code : 03

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12373946

  • £173.00 / Pack of 5


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Can be coiled around flasks, threaded through handles or looped around bottle necks
  • Coated with chemical resistant red Vikem vinyl
  • The 30cm (12 in.) length and flexibility make these weights very versatile

Specifications

Specifications

No
1
Accessory
0.8cm
176g
Videos
Documentation

Technical Documents