Learn More
Bel-Art™ SP Scienceware™ Vacuum Pressure Indicator
Description
- Thermoplastic elastomer collapses with negative vacuum pressure in desiccator and expands when vacuum is absent
- Fits onto tapered end of stopcock
Specifications
Specifications
| Product Type | Vacuum Pressure Indicator |
| For Use With (Application) | Thermoplastic elastomer bellows collapses when under negative pressure (vacuum), expands when under ambient atmospheric pressure, indicates the desiccator vacuum |
| For Use With (Equipment) | Fits on to tapered end of stopcock |