missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.)
- Chemical-resistant polypropylene for use with a wide range of liquids or gases
- FDA-grade for direct contact with food, drugs, and cosmetics
- Autoclavable
Specifications
Specifications
| Certifications/Compliance | FDA |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | 9.5mm |
| Shape | T |
| Color | Natural |
| Product Type | Tubing Connector |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?