missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Super Biohazard Disposal Bags

Protects against dangerous tears and leaks. SP Scienceware™ Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.

Brand:  Bel-Art™ F13165-3748

Additional Details : Weight : 10.20600kg

 View more versions of this product

Product Code. 17692085

  • £625.00 / Pack of 100
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization
Specifications

Specifications

Biohazard Disposal Bag
Orange Red
Autoclavable
Extra-strength, high-impact, high-temperature bags reduce autoclave time
Polymer
37 x 48 in. (94 x 122 cm)
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
2 mil
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers