missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Stir and Add™ cuvette Mixer
Description
- Up and down motion in cell provides complete mixing
- Shaft diameter 3.17 mm (0.125 in.)
- Straight handle
- Used to add enzymes and other materials to solution in spectrophotometer cells.
Specifications
Specifications
| Description | Cell type: Semi-micro; Straight handle |
| Diameter (English) | 0.125 in. |
| For Use With (Application) | Used to add enzymes and other materials to solution in spectrophotometer cells, this tool is used when reactions are performed directly into cuvettes, to mix added reagents |
| Length (English) | 5.1 in. |
| Diameter (Metric) | 3.17 mm |
| Length (Metric) | 13 cm |
| Material | Acrylic |
| Quantity | 36/Cs. |
| Shape | Cylinder |
| Type | Stirring Paddle |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?