Learn More
Bel Art™ ProCulture™ Stak-A-Tray™ System
Product Code. 17692215 Shop All SP Scienceware Products
missing translation for 'orderingAttributeHoverText'
Product Type:
Large Tray Holder
Small Tray Holder
Holds:
Microplates (Standard Size Flat)
Sample Containers
missing translation for 'unitSize'
Each
Product Code. 17692215
missing translation for 'mfr': Bel Art™ H186100470
Only null left
Add to basket
Add to basket
Maximize incubator, cold room and bench space.
  • Keeps trays, flasks and dishes level and stable for even settling of tissue culture cells
  • Racks interlock when stacked, to a maximum of 5 racks high (allows up to 20 levels of trays)
  • Each rack tier holds one large tray (Mfr. No. 18610-1440), 35.6 x 35.6cm, or two smaller trays (Mfr. No. 18610-0470), 17.8 x 35.6cm
  • Perforated trays allow air, gas and water vapor circulation
  • Racks and trays sold separately
  • Stainless-steel storage system is autoclavable
Rack, 2-Tier (Mfr. No. 18610-0420)
  • 5cm clearance between trays, ideal for storing larger culture flasks
Rack, 4-Tier (Mfr. No. 18610-0440)
  • 2.4cm clearance between trays, for storing Petri dishes or smaller culture flasks
Specifications
Product Type Small Tray Holder
Array 12 x 1
Autoclavable Autoclavable
Color Silver
Holds Sample Containers
Material Stainless Steel
Positions 12
Shape Rectangular
Shelves None
Stackable Yes
Sterility Non-sterile
Width (Metric) 178 mm
Height (Metric) 12.5 mm