missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Bottles, Jars and Jugs
- Bottles
- Wash Bottles
- Bel-Art™ Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
Bel-Art™ Right-to-Know Safety-Vented Wash Bottles, GHS Labeling
Product Code. 15569991
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Assorted
Deionized Water
Dichloromethane
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Isotonic Saline
Machine Oil
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite (Bleach)
Toluene
Water
Color:
Assorted
Blue
Green
Natural
Red
Yellow
Quantity:
2/Pk.
4/Pk.
Unit Size:
Pack of 2
Pack of 4
22 product options available for selection
| Product Code. | Capacity (Metric) | Chemical Label | Color | Quantity | unitSize |
|---|---|---|---|---|---|
| 15569991 | 1000 mL | Sodium Hypochlorite (Bleach) | Yellow | 2/Pk. | Pack of 2 |
| 15579971 | 500 mL | Acetone | Red | 4/Pk. | Pack of 4 |
| 15539981 | 500 mL | Machine Oil | Natural | 4/Pk. | Pack of 4 |
| 15599971 | 500 mL | Deionized Water | Blue | 4/Pk. | Pack of 4 |
| 15519981 | 500 mL | Ethyl Acetate | Green | 4/Pk. | Pack of 4 |
| 15589981 | 500 mL | Toluene | Red | 4/Pk. | Pack of 4 |
| 15589971 | 500 mL | Dichloromethane | Yellow | 4/Pk. | Pack of 4 |
| 15509981 | 500 mL | Distilled Water | Blue | 4/Pk. | Pack of 4 |
| 15529981 | 500 mL | Isopropanol | Yellow | 4/Pk. | Pack of 4 |
| 15569981 | 500 mL | Isotonic Saline | Natural | 4/Pk. | Pack of 4 |
| 15549981 | 500 mL | Methanol | Green | 4/Pk. | Pack of 4 |
| 15579981 | 500 mL | Sodium Hypochlorite (Bleach) | Yellow | 4/Pk. | Pack of 4 |
| 15599981 | 500 mL | Water | Blue | 4/Pk. | Pack of 4 |
| 15509991 | 500 mL | Ethanol | Natural | 4/Pk. | Pack of 4 |
| 15519991 | 500 mL | Assorted | Assorted | 4/Pk. | Pack of 4 |
| 15559981 | 500 mL | Methyl Ethyl Ketone | Green | 4/Pk. | Pack of 4 |
| 15559991 | 1000 mL | Methanol | Green | 2/Pk. | Pack of 2 |
| 15529991 | 1000 mL | Acetone | Red | 2/Pk. | Pack of 2 |
| 15549991 | 1000 mL | Isopropanol | Yellow | 2/Pk. | Pack of 2 |
| 15539991 | 1000 mL | Distilled Water | Blue | 2/Pk. | Pack of 2 |
| 15579991 | 1000 mL | Ethanol | Natural | 2/Pk. | Pack of 2 |
| 15589991 | 1000 mL | Assorted | Assorted | 4/Pk. | Pack of 4 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position
- Wide mouth 53 mm (2 in.) for easy refilling
- Translucent LDPE bottles with color-coded caps for easy identification
- Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
- Methyl Ethyl Ketone and Toluene do not include vented valve
- Assortment is 4 per bag, includes one each: Acetone, Isopropanol, Bleach, and Ethanol
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
- Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to Workplace Labels
Specifications
Specifications
| Capacity | 32 oz., 1000 mL |
| Capacity (Metric) | 1000 mL |
| Capacity (English) | 32 oz. |
| Color | Yellow |
| Material | LDPE |
| Quantity | 2/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Mouth | Wide, Threaded |
| Shape | Cylindrical |
| Product Type | Safety-vented Wash Bottle |
| Chemical Label | Sodium Hypochlorite (Bleach) |
| Autoclavable | Not Autoclavable |
| Show More Show Less |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Hittar du en möjlighet till förbättring?