missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ SP Scienceware™ Reusable Desiccant Cartridges

Código de producto. 11317194
Change view
Click to view available options
Diameter (Metric) Outer:
11.4 cm
19.7 cm
23.8 cm
Tamaño de la unidad:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Diameter (Metric) Outer unitSize
11317194 19.7 cm Each
10709475 11.4 cm Each
10555044 23.8 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11317194 Proveedor Bel Art™ N.º de proveedor F420450200

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads. Bel Art SP Scienceware™ Reusable Desiccant Cartridges can also be used with other plastic desiccators of similar size.

  • Desiccant changes color from blue when dry to pink when moist Saturated gel can be regenerated in a 150°C (300°F) vented oven until blue color returns
  • Cartridge can be regenerated repeatedly without degradation
  • Desiccant beads contained in a durable, polyester felt bag with a perforated Mylar°Cover Cartridges are individually sealed in moisture proof pouches ready for immediate use

Información de órdenes

Fits Bel-Art Space Saver™ desiccators in addition to other desiccators of similar size.

Especificaciones

Diameter (Metric) Outer 19.7 cm
Product Type Scienceware™ Reusable Desiccant Cartridge
Material Polyester
For Use With (Application) Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads, for 200mm dia. desiccators
Weight (English) 1.49 lb.
Weight (Metric) 0.68 kg
Diameter (English) Outer 7.75 in.
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Desiccant in a Cartridge
Palabra de advertencia
  • Danger
Clasificación
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Consejos de prudencia
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P501-Dispose of contents/container to .
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.