missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ SP Scienceware™ Reusable Desiccant Cartridges

Product Code. 10709475
Change view
Click to view available options
Diameter (Metric) Outer:
11.4 cm
19.7 cm
23.8 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
10709475 11.4 cm Each
10555044 23.8 cm Each
11317194 19.7 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10709475 Supplier Bel Art™ Supplier No. F420450160

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads. Bel Art SP Scienceware™ Reusable Desiccant Cartridges can also be used with other plastic desiccators of similar size.

  • Desiccant changes color from blue when dry to pink when moist Saturated gel can be regenerated in a 150°C (300°F) vented oven until blue color returns
  • Cartridge can be regenerated repeatedly without degradation
  • Desiccant beads contained in a durable, polyester felt bag with a perforated Mylar°Cover Cartridges are individually sealed in moisture proof pouches ready for immediate use

Order Info

Fits Bel-Art Space Saver™ desiccators in addition to other desiccators of similar size.

Specifications

Diameter (Metric) Outer 11.4 cm
Material Polyester
For Use With (Application) Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads, for 160mm dia. desiccators
Weight (English) 0.5 lb.
Weight (Metric) 0.23 kg
Diameter (English) Outer 4.5 in.
Product Type Reusable Desiccant Cartridge
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Desiccant in a Cartridge
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains cobalt dichloride. May produce an allergic reaction.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.