missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ SP Scienceware™ Reusable Desiccant Cartridges

Product Code. 10555044 Shop All SP Scienceware Products
Click to view available options
Diameter (Metric) Outer:
11.4 cm
19.7 cm
23.8 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10555044

Brand: Bel Art™ F420450250

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads. Bel Art SP Scienceware™ Reusable Desiccant Cartridges can also be used with other plastic desiccators of similar size.

  • Desiccant changes color from blue when dry to pink when moist Saturated gel can be regenerated in a 150°C (300°F) vented oven until blue color returns
  • Cartridge can be regenerated repeatedly without degradation
  • Desiccant beads contained in a durable, polyester felt bag with a perforated Mylar°Cover Cartridges are individually sealed in moisture proof pouches ready for immediate use

Order Info

Fits Bel-Art Space Saver™ desiccators in addition to other desiccators of similar size.

Specifications

Diameter (Metric) Outer 23.8 cm
Product Type Scienceware™ Reusable Desiccant Cartridge
Material Polyester
For Use With (Application) Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads, for 250mm dia. desiccators
Weight (English) 2.99 lb.
Weight (Metric) 1.36 kg
Diameter (English) Outer 9.37 in.
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Desiccant in a Cartridge
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.