Learn More
Bel-Art™ Replacement Chucks for Pipette Pump™ Pipettors
Product Code. 15338740 Shop All SP Scienceware Products
Click to view available options
For Use With (Equipment):
10mL pipette pump pipettor
2mL pump pipettor
Unit Size:
Each
Product Code. 15338740
Brand: Bel-Art™ H937890080
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Grips pipets securely. Bel-Art™ Replacement Chucks are silicone rubber grips used with Bel-Art™ Pipette Pumps for safe, accurate and trouble-free pipetting with simple, one-handed operation.
  • For use with Pipette Pump pipettor with thumbwheel and plunger aspiration
  • Pipettes fit smoothly into these flexible tapered chucks
  • Replacement chucks to fit various sizes/colors of Pipette Pumps in case of loss or damage
  • Silicone rubber is resistant to acids and alkalis
  • Disassemble parts for cleaning; chuck should not be autoclaved
Specifications
Autoclavable Not Autoclavable
For Use With Pipette Pump Pipettors
Material Silicone Rubber
Type Chuck
For Use With (Equipment) 10mL pipette pump pipettor