missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Poxygrid™ Wire Test Tube Racks: 10-13mm

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

Brand:  Bel-Art™ F18753-0001

Code : NEW

Additional Details : Weight : 0.30480kg

 View more versions of this product

Product Code. 15560012

  • £39.25 / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable

Specifications

Specifications

Test Tube Rack
Blue
Epoxy Coated Steel
8 x 6.5 x 2.5 in. (20.5 x 16.4 x 6.4cm)
8.07 in.
6.45 in.
2.51 in.
Autoclavable
Tubes (10 to 13 mm)
108
20.5 cm
16.4 cm
6.5 cm
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers